The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005481.2 | NP_005472.2 | 1 - 2 | 132 | 62 |
XM_017026120.1 | XP_016881609.1 | 1 - 2 | 127 | 62 | |
XM_017026121.1 | XP_016881610.1 | 1 - 2 | 142 | 62 | |
GenBank mRNA | AF106934.1 | - | 1 - 2 | 119 | 62 |
AU098434.1 | - | 1 - 2 | 132 | 62 | |
BC004554.1 | - | 1 - 2 | 113 | 62 | |
BC011841.1 | - | 1 - 2 | 113 | 62 | |
BC017282.2 | - | 1 - 2 | 107 | 62 | |
BI461485.1 | - | 1 - 2 | 134 | 62 |