Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED16
Entrez Gene ID
10025
Gene Name
mediator complex subunit 16
Gene Aliases
DRIP92, THRAP5, TRAP95
Chromosome Location
Chr.19: 867961 - 893228 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005481.2 NP_005472.2 6 - 7 1140 77
XM_017026120.1 XP_016881609.1 6 - 7 1135 77
XM_017026121.1 XP_016881610.1 5 - 6 980 77
GenBank mRNA AF106934.1 - 6 - 7 1127 77
AF121228.1 - 5 - 6 989 77
BC004554.1 - 6 - 7 1121 77
BC007853.2 - 2 - 3 250 77
BC011841.1 - 6 - 7 1121 77
BC017282.2 - 6 - 7 1115 77
EU392449.1 - 5 - 6 1006 77
EU392451.1 - 4 - 5 568 77

View Full Product Details