The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000826.3 | NP_000817.2 | 5 - 6 | 1174 | 126 |
NM_001083619.1 | NP_001077088.1 | 5 - 6 | 1174 | 126 | |
NM_001083620.1 | NP_001077089.1 | 5 - 6 | 685 | 126 | |
XM_017008113.1 | XP_016863602.1 | 5 - 6 | 925 | 126 | |
XM_017008114.1 | XP_016863603.1 | 5 - 6 | 1083 | 126 | |
XM_017008115.1 | XP_016863604.1 | 5 - 6 | 646 | 126 | |
XM_017008116.1 | XP_016863605.1 | 5 - 6 | 870 | 126 | |
XM_017008117.1 | XP_016863606.1 | 5 - 6 | 928 | 126 | |
GenBank mRNA | AB209567.1 | - | 5 - 6 | 1003 | 126 |
AK294458.1 | - | 5 - 6 | 851 | 126 | |
AK308775.1 | - | 5 - 6 | 928 | 126 | |
AK309067.1 | - | 5 - 6 | 698 | 126 | |
AL832605.3 | - | 5 - 6 | 1034 | 126 | |
BC010574.1 | - | 5 - 6 | 994 | 126 | |
BC028736.1 | - | 5 - 6 | 663 | 126 |