The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000826.3 | NP_000817.2 | 8 - 9 | 1621 | 153 |
NM_001083619.1 | NP_001077088.1 | 8 - 9 | 1621 | 153 | |
NM_001083620.1 | NP_001077089.1 | 8 - 9 | 1132 | 153 | |
XM_017008113.1 | XP_016863602.1 | 8 - 9 | 1372 | 153 | |
XM_017008114.1 | XP_016863603.1 | 8 - 9 | 1530 | 153 | |
XM_017008115.1 | XP_016863604.1 | 8 - 9 | 1093 | 153 | |
XM_017008116.1 | XP_016863605.1 | 8 - 9 | 1317 | 153 | |
XM_017008117.1 | XP_016863606.1 | 8 - 9 | 1375 | 153 | |
GenBank mRNA | AB209567.1 | - | 8 - 9 | 1450 | 153 |
AK294458.1 | - | 8 - 9 | 1298 | 153 | |
AK308775.1 | - | 8 - 9 | 1375 | 153 | |
AK309067.1 | - | 8 - 9 | 1145 | 153 | |
AL832605.3 | - | 8 - 9 | 1481 | 153 | |
BC010574.1 | - | 8 - 9 | 1441 | 153 | |
BC028736.1 | - | 8 - 9 | 1110 | 153 | |
L20814.1 | - | 8 - 9 | 1322 | 153 |