Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
Chromosome Location
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001670.2 NP_001661.1 19 - 20 3173 69
XM_005261243.3 XP_005261300.1 18 - 19 3171 69
XM_006724247.3 XP_006724310.1 17 - 18 2833 69
XM_006724248.3 XP_006724311.1 16 - 17 2814 69
GenBank mRNA BC137436.1 - 18 - 19 2951 69
U51269.1 - 19 - 20 3174 69

View Full Product Details