The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003277.3 | NP_003268.2 | 1 - 2 | 184 | 55 |
GenBank mRNA | CB993999.1 | - | - | 211 | 55 |
DB023636.1 | - | 1 - 2 | 184 | 55 |