The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206864.1 | NP_001193793.1 | 4 - 5 | 554 | 133 |
NM_018571.5 | NP_061041.2 | 4 - 5 | 554 | 133 | |
XM_005246669.1 | XP_005246726.1 | 4 - 5 | 508 | 133 | |
GenBank mRNA | AB038950.1 | - | 4 - 5 | 430 | 133 |
AK027637.1 | - | 4 - 5 | 554 | 133 | |
AK290457.1 | - | 5 - 6 | 418 | 133 | |
AY093697.1 | - | 4 - 5 | 442 | 133 | |
BC008302.1 | - | 4 - 5 | 456 | 133 | |
BC090871.1 | - | 4 - 5 | 427 | 133 |