Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSM12
Entrez Gene ID
124801
Gene Name
LSM12 homolog
Gene Aliases
PNAS-135
Chromosome Location
Chr.17: 44034635 - 44067619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152344.3 NP_689557.1 5 - 6 809 89
GenBank mRNA AK055218.1 - 4 - 5 537 89
AL832076.1 - 1 - 1 497 89
BC016791.2 - 1 - 1 310 89
BC044587.1 - 4 - 5 537 89

View Full Product Details