The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004559.4 | NP_004550.2 | 1 - 2 | 494 | 114 |
GenBank mRNA | BC000064.2 | - | 1 - 2 | 281 | 114 |
BC002411.2 | - | 1 - 2 | 277 | 114 | |
BC010430.1 | - | 1 - 2 | 276 | 114 | |
BC015208.1 | - | 1 - 2 | 226 | 114 | |
BC018393.1 | - | 1 - 2 | 261 | 114 | |
BC038384.1 | - | 1 - 2 | 283 | 114 | |
BC065571.1 | - | 1 - 2 | 301 | 114 | |
BC070084.1 | - | 1 - 2 | 301 | 114 | |
BC071708.1 | - | 1 - 2 | 286 | 114 | |
BC072014.1 | - | 1 - 2 | 286 | 114 | |
BC090038.1 | - | 1 - 2 | 286 | 114 | |
BC098435.1 | - | 1 - 2 | 292 | 114 | |
BC106045.1 | - | 1 - 2 | 288 | 114 | |
CN353145.1 | - | 1 - 2 | 343 | 114 | |
J03827.1 | - | 1 - 2 | 289 | 114 | |
L28809.1 | - | 1 - 2 | 244 | 114 | |
M24070.1 | - | 1 - 2 | 283 | 114 |