Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNORD17
Entrez Gene ID
692086
Gene Name
small nucleolar RNA, C/D box 17
Gene Aliases
HBI-43
Chromosome Location
Chr.20: 17962710 - 17962946 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003045.1 1 - 1 143 127

View Full Product Details