Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNORD1A
Entrez Gene ID
677848
Gene Name
small nucleolar RNA, C/D box 1A
Gene Aliases
R38A, snR38A
Chromosome Location
Chr.17: 76561633 - 76561706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_004395.1 1 - 1 38 69


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD653062.1 - 2 477 Chr. 17 - 76557822 - 76562048 [+] on Build GRCh38 69
GenBankmRNA DA274423.1 - 2 475 Chr. 17 - 76557766 - 76561751 [+] on Build GRCh38 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA host gene 16 SNHG16 ncRAN Sense Chr. - 76557764 - 76565348 on Build GRCh38
Downstream ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 ST6GALNAC2 SAITL1, SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM 3.7kb Anti-sense Chr. - 76565379 - 76586066 on Build GRCh38

View Full Product Details