The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_022014.1 | 4 - 5 | 581 | 65 | |
GenBank mRNA | AY271964.1 | - | 4 - 5 | 567 | 65 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 30 member 4 | SLC30A4 | ZNT4, znT-4 | -12.4kb | Anti-sense | Chr. - 45479613 - 45523685 on Build GRCh38 |
Downstream | biogenesis of lysosomal organelles complex 1 subunit 6 | BLOC1S6 | BLOS6, HPS9, PA, PALLID, PLDN | 40.1kb | Sense | Chr. - 45587123 - 45609716 on Build GRCh38 |