Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERICH4
Entrez Gene ID
100170765
Gene Name
glutamate rich 4
Gene Aliases
C19orf69
Chromosome Location
Chr.19: 41442426 - 41444765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130514.1 NP_001123986.1 1 - 2 184 105

View Full Product Details