Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DSCR9
- Entrez Gene ID
-
257203
- Gene Name
- Down syndrome critical region 9 (non-protein coding)
- Gene Aliases
-
NCRNA00038
- Chromosome Location
-
Chr.21: 37208503 - 37221736 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026719.2 |
|
2 - 3 |
149 |
90 |
GenBank mRNA |
AB212286.1 |
- |
2 - 3 |
149 |
90 |
AB212290.1 |
- |
1 - 2 |
107 |
90 |
BC066653.1 |
- |
2 - 3 |
149 |
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ223675.1 |
- |
|
148 |
|
Chr. - - [-] on Build |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tetratricopeptide repeat domain 3 |
TTC3 |
DCRR1, RNF105, TPRDIII |
-5.5kb
|
Sense |
Chr. - 37073184 - 37203118 on Build GRCh38 |
Downstream |
DSCR3 arrestin fold containing |
DSCR3 |
DCRA, DSCRA |
1.7kb
|
Anti-sense |
Chr. - 37223425 - 37268124 on Build GRCh38 |
View Full Product Details