Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HPSE2
- Entrez Gene ID
-
60495
- Gene Name
- heparanase 2 (inactive)
- Gene Aliases
-
HPA2, HPR2, UFS, UFS1
- Chromosome Location
-
Chr.10: 98457077 - 99235875 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021746.1 |
- |
1 - 1 |
1251 |
99 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AU145466.1 |
- |
1 |
|
|
Chr. 10 - 98965908 - 98966428 [+] on Build GRCh38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 6507 |
MIR6507 |
hsa-mir-6507 |
-41.3kb
|
Sense |
Chr. - 98924499 - 98924568 on Build GRCh38 |
Downstream |
cyclin and CBS domain divalent metal cation transport mediator 1 |
CNNM1 |
ACDP1, CLP-1 |
361.5kb
|
Anti-sense |
Chr. - 99329099 - 99394330 on Build GRCh38 |
View Full Product Details