The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 - 3 | 8346 | 95 |
NM_001308340.1 | NP_001295269.1 | 3 - 3 | 4594 | 95 | |
XM_005268665.4 | XP_005268722.1 | 3 - 3 | 8127 | 95 | |
GenBank mRNA | AK123102.1 | - | 1 - 1 | 1173 | 95 |