The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018016.2 | NP_001018016.1 | 1 - 2 | 136 | 76 |
NM_001044392.2 | NP_001037857.1 | 1 - 2 | 136 | 76 | |
NM_001204286.1 | NP_001191215.1 | 1 - 2 | 136 | 76 | |
NM_001204287.1 | NP_001191216.1 | 1 - 2 | 136 | 76 | |
NM_001204288.1 | NP_001191217.1 | 1 - 2 | 136 | 76 | |
NM_001204289.1 | NP_001191218.1 | 1 - 2 | 136 | 76 | |
NM_001204291.1 | NP_001191220.1 | 1 - 2 | 136 | 76 | |
NM_001204292.1 | NP_001191221.1 | 1 - 2 | 136 | 76 | |
NM_001204295.1 | NP_001191224.1 | 1 - 2 | 136 | 76 | |
NM_001204296.1 | NP_001191225.1 | 1 - 2 | 136 | 76 | |
NM_001204297.1 | NP_001191226.1 | 1 - 2 | 136 | 76 | |
GenBank mRNA | AY327584.1 | - | 1 - 2 | 88 | 76 |
AY327586.1 | - | 1 - 2 | 88 | 76 | |
BC120975.2 | - | 1 - 2 | 113 | 76 | |
EF583642.1 | - | 1 - 2 | 114 | 76 | |
EF583644.1 | - | 1 - 2 | 76 | ||
EF583646.1 | - | 1 - 2 | 114 | 76 | |
EF583647.1 | - | 1 - 2 | 114 | 76 | |
EF583648.1 | - | 1 - 2 | 114 | 76 | |
EF583649.1 | - | 1 - 2 | 114 | 76 | |
EF583652.1 | - | 1 - 2 | 114 | 76 | |
EF583653.1 | - | 1 - 2 | 114 | 76 | |
EF583655.1 | - | 1 - 2 | 114 | 76 | |
EF583656.1 | - | 1 - 2 | 114 | 76 | |
EF670708.1 | - | 1 - 2 | 114 | 76 | |
EF670709.1 | - | 1 - 2 | 114 | 76 | |
EF670713.1 | - | 1 - 2 | 114 | 76 | |
EF670716.1 | - | 1 - 2 | 114 | 76 | |
EF670717.1 | - | 1 - 2 | 114 | 76 | |
EF670718.1 | - | 1 - 2 | 114 | 76 | |
FJ226035.1 | - | 1 - 2 | 114 | 76 | |
FJ226040.1 | - | 1 - 2 | 114 | 76 | |
FJ226041.1 | - | 1 - 2 | 114 | 76 | |
FJ226049.1 | - | 1 - 2 | 114 | 76 | |
FJ226051.1 | - | 1 - 2 | 114 | 76 | |
FJ226053.1 | - | 1 - 2 | 114 | 76 | |
FJ226055.1 | - | 1 - 2 | 114 | 76 | |
M31823.1 | - | 1 - 2 | 130 | 76 |