Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- MUC8
- Entrez Gene ID
-
100129528
- Gene Name
- mucin 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 132471092 - 132476638 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U14383.1 |
- |
1 - 1 |
122 |
88 |
View Full Product Details