Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRK-AS1
Entrez Gene ID
100873954
Gene Name
SNRK antisense RNA 1
Gene Aliases
-
Chromosome Location
Chr.3: 43346919 - 43351962 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_046757.1 2 - 3 420 69


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF508610.1 - 1 - 2 Chr. 3 - 43349644 - 43351959 [+] on Build GRCh38 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-) POMGNT2 AGO61, C3orf39, GTDC2, MDDGA8 -240.8kb Sense Chr. - 43079229 - 43106092 on Build GRCh38
Downstream anoctamin 10 ANO10 SCAR10, TMEM16K 14.9kb Sense Chr. - 43366324 - 43691594 on Build GRCh38

View Full Product Details