The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002479.5 | NP_002470.2 | 2 - 3 | 631 | 111 |
GenBank mRNA | BC053899.1 | - | 2 - 3 | 631 | 111 |
BT007233.1 | - | 2 - 3 | 554 | 111 | |
X17651.1 | - | 2 - 3 | 605 | 111 |