Gene Transcripts
- Gene Symbol
- Rrp36
- Entrez Gene ID
- 224823
- Gene Name
- ribosomal RNA processing 36 homolog (S. cerevisiae)
- Gene Aliases
- Chromosome Location
- Chr. 17: 46667442 - 46674451 on Build GRCm38
- UniGene ID
- Mm.45101
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144857.1 |
NP_659106.1
|
2, 3 |
209 |
|
GenBank mRNA |
BC011248.1 |
- |
2, 3 |
209 |
|
View Full Product Details