Gene Transcripts

Gene Symbol
Slc23a3
Entrez Gene ID
22626
Gene Name
solute carrier family 23 (nucleobase transporters), member 3
Gene Aliases
SVCT3, Yspl1
Chromosome Location
Chr. 1: 75125541 - 75135223 on Build GRCm38
UniGene ID
Mm.68157
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194333.3 NP_919314.2 10 1389
GenBank mRNA BC117790.1 - 10 1376
BC117791.1 - 10 1376 40092153
U25739.1 - 10 1401

View Full Product Details