Gene Transcripts

Gene Symbol
Zdhhc5
Entrez Gene ID
228136
Gene Name
zinc finger, DHHC domain containing 5
Gene Aliases
1110032A17Rik, AI451382, Zisp
Chromosome Location
Chr. 2: 84687920 - 84715164 on Build GRCm38
UniGene ID
Mm.288508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144887.4 NP_659136.1 12 3252
GenBank mRNA AK028737.1 - 9 1818
AK075641.1 - 2 598
AK082513.1 - 12 3177
AK145086.1 - 12 3180
AK173251.1 - 12 3207
AY894891.1 - 12 3244
BC020051.1 - 11 2222 3987696
BC027047.1 - 8 1486
BC065155.1 - 12 3122 5706087

View Full Product Details