Gene Transcripts
- Gene Symbol
- Tbc1d17
- Entrez Gene ID
- 233204
- Gene Name
- TBC1 domain family, member 17
- Gene Aliases
- BC017607
- Chromosome Location
- Chr. 7: 44840776 - 44849079 on Build GRCm38
- UniGene ID
- Mm.27567
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042655.1 |
NP_001036120.1
|
1, 2 |
228 |
|
GenBank mRNA |
AK039566.1 |
- |
1, 2 |
43 |
|
AK046883.1 |
- |
1, 2 |
61 |
|
AK081318.1 |
- |
1, 2 |
40 |
|
AK083153.1 |
- |
1, 2 |
45 |
|
AK087754.1 |
- |
|
53 |
|
AK136705.1 |
- |
1, 2 |
53 |
|
AK149917.1 |
- |
1, 2 |
40 |
|
AK153732.1 |
- |
1, 2 |
40 |
|
AK170821.1 |
- |
1, 2 |
40 |
|
BC017607.1 |
- |
1, 2 |
228 |
4507589 |
BC035449.1 |
- |
1, 2 |
27 |
|
View Full Product Details