Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010827.2 | NP_034957.1 | 1 | 348 | |
GenBank mRNA | AB221639.1 | - | 1 | 130 | |
AF087035.1 | - | 1 | 348 | ||
AF108216.1 | - | 1 | 130 | ||
AK138917.1 | - | 1 | 375 | ||
BC103592.1 | - | 1 | 255 | 40046034 | |
BC103593.1 | - | 1 | 255 | 40046035 | |
BC103594.1 | - | 1 | 251 | 40046036 | |
BC103623.1 | - | 1 | 255 | 40046489 |