Gene Transcripts

Gene Symbol
Acy3
Entrez Gene ID
71670
Gene Name
aspartoacylase (aminoacylase) 3
Gene Aliases
0610006H10Rik, AA3, AAIII, AW107362, Acy-3, HCBP1
Chromosome Location
Chr. 19: 3986570 - 3990007 on Build GRCm38
UniGene ID
Mm.29735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF356878.1 - 5 681
AF356879.1 - 4 540
AF375479.1 - 3 492
AK002247.1 - 5 680
AK143782.1 - 5 683
AY169234.1 - 5 680
BC010795.1 - 3 901 4221711

View Full Product Details