Gene Transcripts

Gene Symbol
Uba7
Entrez Gene ID
74153
Gene Name
ubiquitin-like modifier activating enzyme 7
Gene Aliases
1300004C08Rik, Ube1l
Chromosome Location
Chr. 9: 107975567 - 107984056 on Build GRCm38
UniGene ID
Mm.277125
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023738.4 NP_076227.1 16 2046
GenBank mRNA AF288694.1 - 11 1308
AK004894.2 - 16 2047
AK054410.1 - 15 2138
AK089235.1 - 15 1939
AK089256.1 - 7 1715
AK129009.1 - 16 1956
AK170421.1 - 15 2124
AK170644.1 - 14 2506
AK172467.1 - 15 2384
BC037693.1 - 14 1820
BC138843.1 - 16 1963 8861865

View Full Product Details