Product Details

SNP ID
rs13482941
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:29327410 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGCAGAGGTCGAGTTGCGGGA[A/G]CCCGAGGCTGAGTCTCCCAAGGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mtch1 PubMed Links

Gene Details

Gene
Mtch1
Gene Name
mitochondrial carrier homolog 1 (C. elegans)
There are no transcripts associated with this gene.

Gene
Pi16
Gene Name
peptidase inhibitor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023734.3 1494 Silent Mutation NP_076223.3

View Full Product Details