Product Details
- SNP ID
-
rs13480875
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:12295807 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCATTTCTCCCTTGCACAGCTAAG[C/T]AAAACCCATGTGTTTGTTATCTACT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cobl
PubMed Links
Gene Details
- Gene
- Cobl
- Gene Name
- cordon-bleu WH2 repeat
View Full Product Details