Product Details

SNP ID
rs4229749
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:53332910 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GATCACTTAAGAAGATGCCGACCCA[C/T]GGTTCACATGAACGCACATCAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Sptlc1 PubMed Links

Gene Details

Gene
Sptlc1
Gene Name
serine palmitoyltransferase, long chain base subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009269.2 2408 UTR 3 NP_033295.2

View Full Product Details