Product Details
- SNP ID
-
rs13483825
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:74207542 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAACTCTTTAAGCCTGTTGGAGC[A/G]TGACTATTGGAGTGAATCCATGAGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tktl1
PubMed Links
Gene Details
- Gene
- Tktl1
- Gene Name
- transketolase-like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031379.2 |
|
Intron |
|
|
NP_113556.2 |
View Full Product Details