Product Details

SNP ID
rs13483014
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:49070207 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGACCTTGAGAGCCTCCGCTCCCT[A/G]CACCTTGACAGCAACCGGCTACCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Lrfn2 PubMed Links

Gene Details

Gene
Lrfn2
Gene Name
leucine rich repeat and fibronectin type III domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027452.3 576 Silent Mutation NP_081728.2

View Full Product Details