Product Details

SNP ID
rs3661835
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:12920189 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACACATGACCAATTTGGATGCCTTT[A/T]TAAATACAAACTGAAGCAGATATTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Slco5a1 PubMed Links

Gene Details

Gene
Slco5a1
Gene Name
solute carrier organic anion transporter family, member 5A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172841.2 Intron NP_766429.2

View Full Product Details