Product Details
- SNP ID
-
rs3710340
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:171938066 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACTACAATTTCAATAATCAGTGC[T/A]GTCATACTCATCTTTGTGTGCTGGA
- Phenotype
-
- Polymorphism
- T/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Slamf6
PubMed Links
Gene Details
- Gene
- Slamf6
- Gene Name
- SLAM family member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030710.2 |
927 |
Silent Mutation |
|
|
NP_109635.1 |
View Full Product Details