Product Details

SNP ID
rs4228165
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:26274577 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGTAACAATAAAAAGCTTACTTAG[C/T]AGATTCATGCAGAGAAATGAAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
L3mbtl3 PubMed Links

Gene Details

Gene
L3mbtl3
Gene Name
l(3)mbt-like 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172787.2 Intron NP_766375.1
Gene
Samd3
Gene Name
sterile alpha motif domain containing 3
There are no transcripts associated with this gene.

View Full Product Details