Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSTD2
Entrez Gene ID
102128745
Gene Name
thiosulfate sulfurtransferase like domain containing 2
Gene Aliases
-
Chromosome Location
Chr.15: 40144639 - 40178418 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581239.2 XP_005581296.1 6 - 7 1296 154

View Full Product Details