Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPSE2
Entrez Gene ID
102138349
Gene Name
heparanase 2 (inactive)
Gene Aliases
-
Chromosome Location
Chr.9: 97286585 - 98087375 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566158.2 XP_005566215.1 11 - 12 1715 69

View Full Product Details