Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPPP2
Entrez Gene ID
101925360
Gene Name
tubulin polymerization promoting protein family member 2
Gene Aliases
-
Chromosome Location
Chr.7: 84582391 - 84584384 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285278.1 NP_001272207.1 3 - 4 478 80
XM_005560742.1 XP_005560799.1 3 - 4 511 80
XM_015453234.1 XP_015308720.1 3 - 4 514 80
GenBank mRNA AB179366.1 - 3 - 4 478 80

View Full Product Details