Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC9
Entrez Gene ID
102126211
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
-
Chromosome Location
Chr.9: 62303672 - 62322005 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565521.2 XP_005565578.1 3 - 4 2248 129
XM_005565523.2 XP_005565580.1 3 - 4 781 129
XM_005565524.2 XP_005565581.1 3 - 4 781 129
XM_005565525.2 XP_005565582.1 3 - 4 781 129

View Full Product Details