Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GM2A
Entrez Gene ID
102143555
Gene Name
GM2 ganglioside activator
Gene Aliases
GM2-AP, SAP-3
Chromosome Location
Chr.6: 150548868 - 150563654 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283562.1 NP_001270491.1 1 - 2 144 91
GenBank mRNA AB083313.2 - 1 - 2 144 91
AB125172.1 - 1 - 2 182 91
AB170659.1 - 1 - 2 115 91

View Full Product Details