Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLAMF9
Entrez Gene ID
102117069
Gene Name
SLAM family member 9
Gene Aliases
-
Chromosome Location
Chr.1: 91718551 - 91721310 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302658.1 NP_001289587.1 3 - 4 781 86
XM_015451946.1 XP_015307432.1 3 - 4 805 86

View Full Product Details