Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102116465
Entrez Gene ID
102116465
Gene Name
aldo-keto reductase family 1 member C2
Gene Aliases
AKR1C2
Chromosome Location
Chr.9: 5264266 - 5276702 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015455492.1 XP_015310978.1 4 - 5 493 83
XM_015455493.1 XP_015310979.1 4 - 5 493 83

View Full Product Details