Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CST5
Entrez Gene ID
102146503
Gene Name
cystatin D
Gene Aliases
-
Chromosome Location
Chr.10: 38435199 - 38439867 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568164.2 XP_005568221.1 2 - 3 454 79
XM_015457730.1 XP_015313216.1 3 - 4 770 79

View Full Product Details