Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102122664
Entrez Gene ID
102122664
Gene Name
alcohol dehydrogenase 1A
Gene Aliases
ADH1A
Chromosome Location
Chr.5: 98548112 - 98562860 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555516.2 XP_005555573.1 7 - 8 1075 95

View Full Product Details