The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290414.1 | NP_001277343.1 | 5 - 6 | 782 | 116 |
NM_001290415.1 | NP_001277344.1 | 3 - 4 | 293 | 116 | |
NM_010200.3 | NP_034330.2 | 3 - 4 | 777 | 116 | |
GenBank mRNA | AF020737.1 | - | 3 - 4 | 410 | 116 |
AK141848.1 | - | 3 - 4 | 293 | 116 | |
BC018238.1 | - | 3 - 4 | 777 | 116 | |
U66202.1 | - | 3 - 4 | 404 | 116 |