Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gria2
Entrez Gene ID
14800
Gene Name
glutamate receptor, ionotropic, AMPA2 (alpha 2)
Gene Aliases
GluA2, GluR-B, Glur-2, Glur2
Chromosome Location
Chr.3: 80684935 - 80803204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039195.1 NP_001034284.1 4 - 5 1071 110
NM_001083806.1 NP_001077275.1 4 - 5 1071 110
NM_013540.2 NP_038568.2 4 - 5 1071 110
GenBank mRNA AB109210.1 - 4 - 5 664 110
AB109211.1 - 4 - 5 664 110
AB111957.1 - 4 - 5 664 110
AK014389.1 - 4 - 5 1116 110
AK043490.1 - 4 - 5 1044 110
AK044574.1 - 4 - 5 1003 110
AK046861.1 - 4 - 5 1071 110
AK086614.1 - 4 - 5 1005 110
AK134572.1 - 4 - 5 1002 110
AK134647.1 - 3 - 4 517 110
AK134772.1 - 3 - 4 642 110
AK141525.1 - 4 - 5 1046 110
AK163941.1 - 4 - 5 1064 110
BC048248.1 - 4 - 5 930 110
BC066193.1 - 4 - 5 859 110
X57498.1 - 4 - 5 866 110

View Full Product Details