Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smtn
Entrez Gene ID
29856
Gene Name
smoothelin
Gene Aliases
smsmo
Chromosome Location
Chr.11: 3517522 - 3539654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159284.1 NP_001152756.1 17 - 18 2471 103
NM_001284427.1 NP_001271356.1 16 - 17 2420 103
NM_001284428.1 NP_001271357.1 17 - 18 2528 103
NM_001284429.1 NP_001271358.1 17 - 18 2473 103
NM_013870.3 NP_038898.2 16 - 17 2420 103
GenBank mRNA AF064236.1 - 16 - 17 2367 103
AJ010305.1 - 16 - 17 2367 103
AK009426.1 - 4 - 5 833 103
AK030932.1 - 16 - 17 2421 103
AK083947.1 - 6 - 7 674 103
AK085412.1 - 17 - 18 2436 103
AK138898.1 - 17 - 18 2486 103
AK160492.1 - 7 - 8 875 103
AK170516.1 - 16 - 17 2380 103
BC010599.1 - 16 - 17 2361 103
BC066192.1 - 17 - 18 2346 103
BC069836.1 - 17 - 18 2346 103
BC069839.1 - 17 - 18 2346 103

View Full Product Details