Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thop1
Entrez Gene ID
50492
Gene Name
thimet oligopeptidase 1
Gene Aliases
AI131655, AI327041, EP24.15
Chromosome Location
Chr.10: 81070083 - 81082360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022653.4 NP_073144.3 12 - 13 2091 97
GenBank mRNA AF314187.1 - 12 - 13 2049 97
AK028609.1 - 12 - 13 2025 97
AK185106.1 - 1 - 2 102 96
BC031175.1 - 4 - 5 602 97
BC031722.1 - 12 - 13 2057 97
BC039651.1 - 4 - 5 602 97

View Full Product Details