The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010120.5 | NP_034250.3 | 2 - 3 | 155 | 87 |
GenBank mRNA | AF026481.1 | - | 1 - 2 | 87 | |
AK009026.1 | - | 2 - 3 | 126 | 87 | |
AK019451.1 | - | 2 - 3 | 156 | 87 | |
AK150651.1 | - | 2 - 3 | 152 | 87 | |
AK150673.1 | - | 2 - 3 | 128 | 87 | |
AK150677.1 | - | 2 - 3 | 128 | 87 | |
AK166368.1 | - | - | 127 | 87 | |
AK167333.1 | - | 2 - 3 | 115 | 87 | |
BC027437.1 | - | 2 - 3 | 117 | 87 |