Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcoln3
Entrez Gene ID
171166
Gene Name
mucolipin 3
Gene Aliases
6720490O21Rik, TRPML3, Va
Chromosome Location
Chr.3: 146093277 - 146141808 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134160.1 NP_598921.1 10 - 11 1324 59
GenBank mRNA AF475086.1 - 10 - 11 1324 59
AK030819.1 - 10 - 11 1444 59
AK033008.1 - 10 - 11 1438 59
AK035029.1 - 10 - 11 1445 59
AY083531.1 - 10 - 11 1322 59
BC106856.2 - 10 - 11 1324 59
BC106857.2 - 10 - 11 1324 59

View Full Product Details